Assay Details
Target Gene Details
Entrez Gene ID: | 130162 |
Gene Name: | clathrin heavy chain linker domain containing 1 |
Gene Aliases: |
C2orf63 |
Location: |
Chr.2:55172547-55232563 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2720078 | Chr.2:53962225 - 55281329 on Build GRCh38 | Deletion | RPL23AP32 SPTBN1 RTN4 CLHC1 EML6 ACYP2 C2orf73 TSPYL6 PSME4 RPS27A MTIF2 |
nsv1010438 | Chr.2:55146426 - 55223008 on Build GRCh38 | Loss | CLHC1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |