Assay Details
Target Gene Details
Entrez Gene ID: | 419 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
ARTC3 |
Location: |
Chr.4:76011180-76112802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ART3 | NM_001130017.2 | NP_001123489.1 | ||
AK225860.1 | ||||
AY543656.1 | AAT10278.1 | |||
AY543657.1 | AAT10279.1 | |||
AY543658.1 | AAT10280.1 | |||
AY543659.1 | AAT10281.1 | |||
DB451386.1 | ||||
U47054.1 | AAB01894.1 |
Target Gene Details
Entrez Gene ID: | 6373 |
Gene Name: | C-X-C motif chemokine ligand 11 |
Gene Aliases: |
H174, I-TAC, IP-9, IP9, SCYB11, SCYB9B, b-R1 |
Location: |
Chr.4:76033682-76036197 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCL11 | NM_001302123.1 | NP_001289052.1 | ||
NM_005409.4 | NP_005400.1 | |||
AF002985.1 | AAC51845.1 | |||
AF030514.1 | AAC39775.1 | |||
AF352781.1 | AAK52900.1 | |||
AL703031.1 | ||||
BC005292.1 | AAH05292.1 | |||
BC012532.1 | AAH12532.1 | |||
BC110986.1 | AAI10987.1 | |||
BP289021.1 | ||||
BT006787.1 | AAP35433.1 | |||
U59286.1 | AAB17374.1 | |||
U66096.1 | AAD38867.1 | |||
Y15220.1 | CAA75510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv27112 | Chr.4:76027416 - 76056627 on Build GRCh38 | Gain | ART3 CXCL11 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |