Assay Details
Target Gene Details
Entrez Gene ID: | 3066 |
Gene Name: | histone deacetylase 2 |
Gene Aliases: |
HD2, RPD3, YAF1 |
Location: |
Chr.6:113936156-113971195 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HDAC2 | NM_001527.3 | 11 | 1448 | NP_001518.3 |
NR_033441.1 | 12 | 1476 | ||
NR_073443.1 | 11 | 1354 | ||
XM_011535788.1 | 12 | 1200 | XP_011534090.1 | |
XM_017010799.1 | 12 | 1422 | XP_016866288.1 | |
AB209190.1 | 5 | 4164 | BAD92427.1 | |
AK092156.1 | 11 | 1354 | ||
AK097376.1 | 12 | 1410 | ||
AK296856.1 | 11 | 1448 | ||
BC031055.1 | 11 | 1412 | AAH31055.2 | |
U31814.1 | 11 | 1365 | AAC50814.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |