Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | NP_001305309.1 | ||
NM_177533.4 | NP_803877.2 | |||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
AK310449.1 | ||||
BC041584.1 | AAH41584.1 | |||
DA249638.1 | 4 | 434 | ||
DA409636.1 | 4 | 497 | ||
DB096906.1 | 4 | 516 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | TEX22 PACS2 MTA1 LOC100507437 LOC102723354 BTBD6 GPR132 BRF1 JAG2 CRIP2 MIR6765 NUDT14 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | TEX22 CRIP1 C14orf80 CRIP2 MIR6765 IGH PACS2 MTA1 LOC105370697 LOC100507437 IGHA2 BTBD6 BRF1 JAG2 TMEM121 NUDT14 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | LOC102723354 GPR132 BRF1 JAG2 MIR6765 NUDT14 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | BRF1 JAG2 MIR6765 NUDT14 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | CEP170B LINC00638 TRMT61A INF2 XRCC3 C14orf79 MIR203A PPP1R13B ASPG LOC101929634 SIVA1 C14orf2 APOPT1 KIF26A KLC1 MIR203B ZFYVE21 LOC105370691 BTBD6 BRF1 ZBTB42 SNORA28 RD3L ADSSL1 NUDT14 LOC105370708 TMEM179 LOC102723354 LINC00637 LOC107984648 GPR132 CDCA4 AHNAK2 CKB MIR6765 MARK3 AKT1 PACS2 LOC105378183 MIR4710 LOC105370687 EIF5 TDRD9 LOC388022 C14orf180 JAG2 BAG5 PLD4 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | LOC102723354 C14orf79 BTBD6 GPR132 CDCA4 BRF1 AHNAK2 JAG2 MIR6765 NUDT14 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGHV3-71 IGHV4-31 CRIP1 MIR4538 C14orf80 ELK2AP IGHG3 LINC00226 IGHA1 KIAA0125 IGH MIR4537 MTA1 LOC105370697 LOC100507437 IGHA2 IGHV3-48 IGHV3-30 IGHG4 IGHV3-23 BTBD6 BRF1 IGHV4-55 TMEM121 IGHD NUDT14 MIR8071-2 TEX22 ADAM6 MIR4539 IGHV3-69-1 MIR4507 LINC00221 CRIP2 MIR6765 PACS2 IGHG1 MIR8071-1 LOC107987211 IGHV1-69 IGHV3-41 JAG2 IGHG2 IGHM |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | LOC102723354 C14orf79 GPR132 CDCA4 BRF1 AHNAK2 JAG2 MIR6765 NUDT14 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |