Assay Details
Target Gene Details
Entrez Gene ID: | 404220 |
Gene Name: | chromosome 6 open reading frame 201 |
Gene Aliases: |
dJ1013A10.5 |
Location: |
Chr.6:4079197-4130788 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf201 | NM_001085401.2 | 1 | 363 | NP_001078870.1 |
NR_104463.1 | 1 | 363 | ||
NR_104464.1 | 1 | 363 | ||
XM_011514613.2 | XP_011512915.1 | |||
XM_017010861.1 | 1 | 372 | XP_016866350.1 | |
BC047663.1 | 1 | 360 | AAH47663.1 | |
BI459567.1 | 1 | 365 |
Target Gene Details
Entrez Gene ID: | 222826 |
Gene Name: | family with sequence similarity 217 member A |
Gene Aliases: |
C6orf146 |
Location: |
Chr.6:4068352-4087344 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM217A | XM_011514415.1 | XP_011512717.1 | ||
XM_017010482.1 | XP_016865971.1 | |||
AK097426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv600860 | Chr.6:4052149 - 4096444 on Build GRCh38 | Loss | FAM217A PRPF4B C6orf201 |