Assay Details
Target Gene Details
Entrez Gene ID: | 65005 |
Gene Name: | mitochondrial ribosomal protein L9 |
Gene Aliases: |
L9mt |
Location: |
Chr.1:151759643-151763916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL9 | NM_001300733.1 | NP_001287662.1 | ||
NM_031420.3 | NP_113608.1 | |||
NR_125331.1 | ||||
AB049636.1 | BAB40841.1 | |||
AK026363.1 | ||||
AK293401.1 | ||||
AK300668.1 | ||||
AK309841.1 | 2 | 343 | ||
AK315459.1 | ||||
BC004517.2 | AAH04517.1 | |||
BG437768.1 | ||||
CF127136.1 |
Target Gene Details
Entrez Gene ID: | 51686 |
Gene Name: | ornithine decarboxylase antizyme 3 |
Gene Aliases: |
AZ3, OAZ-t, TISP15 |
Location: |
Chr.1:151762969-151771330 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OAZ3 | NM_001134939.1 | NP_001128411.1 | ||
BC073949.1 | AAH73949.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |