Assay Details
Target Gene Details
Entrez Gene ID: | 9049 |
Gene Name: | aryl hydrocarbon receptor interacting protein |
Gene Aliases: |
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2 |
Location: |
Chr.11:67483029-67491108 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIP | NM_001302959.1 | 4 | 620 | NP_001289888.1 |
NM_001302960.1 | 4 | 654 | NP_001289889.1 | |
NM_003977.3 | 4 | 654 | NP_003968.3 | |
BC104797.1 | 4 | 556 | AAI04798.1 | |
BC104827.1 | 4 | 556 | AAI04828.1 | |
BG821308.1 | 4 | 649 | ||
BQ054206.1 | 4 | 620 | ||
BQ420398.1 | 3 | 380 | ||
BU689235.1 | 3 | 598 | ||
CN480874.1 | 3 | 602 | ||
U31913.1 | 4 | 533 | AAB39923.1 | |
U78521.1 | 4 | 641 | AAB59004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | CDK2AP2 RPS6KB2 CABP4 PTPRCAP GPR152 PITPNM1 MIR6752 CORO1B CARNS1 AIP TMEM134 |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication | ALDH3B2 CDK2AP2 GSTP1 CABP2 PITPNM1 AIP DOC2GP TMEM134 NDUFV1 FAM86C2P ACY3 C11orf72 NUDT8 MIR6752 TBX10 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | CDK2AP2 PPP1CA RAD9A TBC1D10C RPS6KB2 CABP4 CABP2 PTPRCAP GPR152 PITPNM1 CORO1B AIP TMEM134 MIR6752 CARNS1 |
nsv368 | Chr.11:67479842 - 67509866 on Build GRCh38 | Insertion | CDK2AP2 PITPNM1 MIR6752 AIP |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | CDK2AP2 PPP1CA RAD9A TBC1D10C RPS6KB2 CABP4 CABP2 PTPRCAP LOC100130987 GPR152 PITPNM1 CORO1B CLCF1 AIP TMEM134 MIR6752 CARNS1 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | CDK2AP2 TBC1D10C RPS6KB2 CABP4 CABP2 PTPRCAP GPR152 PITPNM1 CORO1B AIP TMEM134 MIR6752 CARNS1 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | CDK2AP2 CABP4 CABP2 GPR152 PITPNM1 MIR6752 CORO1B AIP TMEM134 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |