Assay Details
Target Gene Details
Entrez Gene ID: | 10580 |
Gene Name: | sorbin and SH3 domain containing 1 |
Gene Aliases: |
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1 |
Location: |
Chr.10:95311773-95561420 on Build GRCh38 |
Assay Gene Location: | Within Exon 36 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3624276 | Chr.10:95309786 - 95356777 on Build GRCh38 | Gain | SORBS1 |
dgv161e214 | Chr.10:95309657 - 95503271 on Build GRCh38 | Gain | SORBS1 |
esv22688 | Chr.10:95096249 - 95314999 on Build GRCh38 | Gain+Loss | SORBS1 PDLIM1 ACSM6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |