Assay Details
Target Gene Details
Entrez Gene ID: | 7163 |
Gene Name: | tumor protein D52 |
Gene Aliases: |
D52, N8L, PC-1, PrLZ, hD52 |
Location: |
Chr.8:80034868-80171659 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPD52 | NM_001025252.2 | 6 | 1208 | NP_001020423.1 |
NM_001025253.2 | 8 | 995 | NP_001020424.1 | |
NM_001287140.1 | 8 | 1277 | NP_001274069.1 | |
NM_001287142.1 | 7 | 1250 | NP_001274071.1 | |
NM_001287143.1 | 7 | 1235 | NP_001274072.1 | |
NM_001287144.1 | 6 | 919 | NP_001274073.1 | |
NM_005079.3 | 6 | 926 | NP_005070.1 | |
NR_105033.1 | 9 | 1347 | ||
NR_105034.1 | 5 | 810 | ||
NR_105035.1 | 7 | 996 | ||
NR_105036.1 | 8 | 940 | ||
NR_105037.1 | 7 | 941 | ||
AF202897.1 | 6 | 1152 | AAO22156.1 | |
AK057075.1 | 8 | 996 | ||
AK223631.1 | 6 | 825 | BAD97351.1 | |
AK295468.1 | 8 | 862 | ||
AK296615.1 | 6 | 930 | ||
AK308983.1 | 8 | 852 | ||
BC018117.1 | 6 | 797 | AAH18117.1 | |
BX640835.1 | 6 | 992 | CAE45908.1 | |
GQ499324.1 | 8 | 1262 | ||
GQ499325.1 | 7 | 1235 | ||
GQ499326.1 | 7 | 1220 | ||
GQ499327.1 | 6 | 1193 | ||
GQ499328.1 | 9 | 1332 | ||
S82081.1 | 6 | 803 | AAB36475.1 | |
U18914.1 | 6 | 801 | AAC50183.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |