Assay Details
Target Gene Details
Entrez Gene ID: | 8501 |
Gene Name: | solute carrier family 43 member 1 |
Gene Aliases: |
LAT3, PB39, POV1, R00504 |
Location: |
Chr.11:57484531-57515954 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC43A1 | NM_001198810.1 | 15 | 1826 | NP_001185739.1 |
NM_003627.5 | 15 | 1997 | NP_003618.1 | |
XM_005274358.4 | 16 | 2137 | XP_005274415.1 | |
XM_011545320.2 | 15 | 2046 | XP_011543622.1 | |
XM_011545321.2 | 13 | 1613 | XP_011543623.1 | |
XM_011545322.1 | 12 | 1462 | XP_011543624.1 | |
XM_017018451.1 | 16 | 2025 | XP_016873940.1 | |
XM_017018452.1 | 14 | 1966 | XP_016873941.1 | |
XM_017018453.1 | 13 | 1803 | XP_016873942.1 | |
AB103033.1 | 15 | 1964 | BAD00152.1 | |
AF045584.1 | 15 | 1784 | AAC33004.1 | |
AK027732.1 | 13 | 1562 | ||
BC001639.2 | 15 | 1826 | AAH01639.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |