Assay Details
Target Gene Details
Entrez Gene ID: | 1796 |
Gene Name: | docking protein 1 |
Gene Aliases: |
P62DOK, pp62 |
Location: |
Chr.2:74549020-74557551 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOK1 | NM_001197260.1 | 5 | 1273 | NP_001184189.1 |
NM_001318866.1 | 5 | 1773 | NP_001305795.1 | |
NM_001318867.1 | 5 | 1818 | NP_001305796.1 | |
NM_001318868.1 | 4 | 1590 | NP_001305797.1 | |
NM_001318869.1 | 4 | 1584 | NP_001305798.1 | |
NM_001381.4 | 5 | 1775 | NP_001372.1 | |
XM_017003528.1 | 3 | 1696 | XP_016859017.1 | |
AF035299.1 | 5 | 1266 | AAB88182.1 | |
AF180527.1 | 4 | 995 | AAF19167.1 | |
AK055944.1 | 5 | 1775 | ||
AK127127.1 | 3 | 1471 | ||
AK127656.1 | 3 | 1471 | ||
AK222883.1 | 5 | 1214 | BAD96603.1 | |
AK296154.1 | 5 | 1228 | ||
AK300097.1 | 4 | 995 | ||
AK314678.1 | 5 | 1195 | ||
BC111055.1 | 4 | 1019 | AAI11056.1 | |
BC114440.1 | 5 | 1201 | ||
BC114497.1 | 4 | 1004 | ||
U70987.1 | 5 | 1202 | AAC51127.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |