Assay Details
Target Gene Details
Entrez Gene ID: | 54798 |
Gene Name: | dachsous cadherin-related 2 |
Gene Aliases: |
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ |
Location: |
Chr.4:154234375-154491778 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCHS2 | NM_017639.3 | 25 | 6678 | NP_060109.2 |
XM_011532045.1 | 20 | 8466 | XP_011530347.1 | |
XM_011532046.2 | 19 | 6122 | XP_011530348.1 | |
XM_011532049.2 | 12 | 3972 | XP_011530351.1 | |
AF169691.1 | 1 | 990 | AAF89688.1 | |
AK000054.1 | 3 | 975 | BAA90911.1 | |
AK095103.1 | 8 | 2015 | ||
AK225200.1 | 3 | 975 | ||
AY354497.1 | 25 | 6678 | AAR10443.1 | |
BX647835.1 | 8 | 1654 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023270 | Chr.4:154225348 - 154311669 on Build GRCh38 | Gain | DCHS2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |