Assay Details
Target Gene Details
Entrez Gene ID: | 115352 |
Gene Name: | Fc receptor like 3 |
Gene Aliases: |
CD307c, FCRH3, IFGP3, IRTA3, SPAP2 |
Location: |
Chr.1:157676481-157700985 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL3 | NM_001320333.1 | NP_001307262.1 | ||
NM_052939.3 | NP_443171.2 | |||
NR_135214.1 | ||||
NR_135215.1 | ||||
NR_135216.1 | ||||
NR_135217.1 | ||||
XM_006711145.1 | XP_006711208.1 | |||
AF416901.1 | AAL13290.1 | |||
AF416902.1 | AAL13291.1 | |||
AF416903.1 | AAL13292.1 | |||
AF416904.1 | AAL13293.1 | |||
AF416905.1 | AAL13294.1 | |||
AF459027.1 | AAL59390.1 | |||
AK098122.1 | ||||
AK301257.1 | ||||
AL833391.1 | 1 | 892 | ||
AY043466.1 | AAK91779.1 | |||
BC028933.1 | AAH28933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520011 | Chr.1:157678308 - 157703838 on Build GRCh38 | Loss | FCRL3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |