Assay Details
Target Gene Details
Entrez Gene ID: | 56063 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
Location: |
Chr.1:32204807-32222371 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM234 | NM_019118.4 | 5 | 951 | NP_061991.3 |
NR_133634.1 | 6 | 1092 | ||
NR_133635.1 | ||||
NR_133636.1 | ||||
XM_011541795.2 | XP_011540097.1 | |||
XM_017001812.1 | 4 | 1244 | XP_016857301.1 | |
XM_017001813.1 | 4 | 1244 | XP_016857302.1 | |
XM_017001814.1 | XP_016857303.1 | |||
XM_017001815.1 | XP_016857304.1 | |||
XM_017001816.1 | XP_016857305.1 | |||
XM_017001817.1 | 6 | 562 | XP_016857306.1 | |
XM_017001818.1 | XP_016857307.1 | |||
XM_017001819.1 | XP_016857308.1 | |||
XM_017001820.1 | XP_016857309.1 | |||
XM_017001821.1 | XP_016857310.1 | |||
XM_017001822.1 | XP_016857311.1 | |||
XM_017001823.1 | XP_016857312.1 | |||
XM_017001824.1 | 6 | 562 | XP_016857313.1 | |
XM_017001825.1 | 5 | 1110 | XP_016857314.1 | |
XM_017001826.1 | 5 | 933 | XP_016857315.1 | |
XM_017001827.1 | XP_016857316.1 | |||
XM_017001828.1 | XP_016857317.1 | |||
XM_017001829.1 | XP_016857318.1 | |||
XM_017001830.1 | XP_016857319.1 | |||
AF258548.1 | AAG23751.1 | |||
AK075260.1 | 2 | 1044 | BAC11504.1 | |
AK295232.1 | ||||
AK312046.1 | ||||
AL512700.1 | 1 | 1375 | ||
AY358586.1 | 4 | 1240 | AAQ88949.1 | |
BC004885.2 | 5 | 905 | AAH04885.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv300 | Chr.1:32206912 - 32241689 on Build GRCh38 | Insertion | DCDC2B TMEM234 IQCC EIF3I MTMR9LP |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |