Assay Details
Target Gene Details
Entrez Gene ID: | 2060 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 |
Gene Aliases: |
AF-1P, AF1P, MLLT5 |
Location: |
Chr.1:51354263-51519354 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS15 | NM_001159969.1 | 13 | 2077 | NP_001153441.1 |
NM_001981.2 | 25 | 2946 | NP_001972.1 | |
XM_005270618.2 | 24 | 2887 | XP_005270675.1 | |
XM_017000615.1 | 25 | 3088 | XP_016856104.1 | |
XM_017000616.1 | 24 | 2884 | XP_016856105.1 | |
XM_017000617.1 | 24 | 2773 | XP_016856106.1 | |
XM_017000618.1 | 23 | 2683 | XP_016856107.1 | |
XM_017000619.1 | 26 | 2970 | XP_016856108.1 | |
XM_017000620.1 | 25 | 3137 | XP_016856109.1 | |
AF052132.1 | 2 | 466 | ||
AK090936.1 | 13 | 1803 | ||
AK294297.1 | 14 | 1926 | ||
AK308128.1 | ||||
BC054006.1 | 24 | 2462 | AAH54006.1 | |
BX647676.1 | 13 | 2058 | ||
U07707.1 | 25 | 2875 | AAA52101.1 | |
Z29064.1 | 25 | 2946 | CAA82305.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |