Assay Details
Target Gene Details
Entrez Gene ID: | 7082 |
Gene Name: | tight junction protein 1 |
Gene Aliases: |
ZO-1 |
Location: |
Chr.15:29699367-29968919 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TJP1 | NM_001301025.1 | 29 | 6626 | NP_001287954.1 |
NM_001301026.1 | 28 | 6401 | NP_001287955.1 | |
NM_003257.4 | 28 | 6802 | NP_003248.3 | |
NM_175610.3 | 27 | 6562 | NP_783297.2 | |
XM_005254617.3 | 29 | 6686 | XP_005254674.2 | |
XM_005254618.3 | 28 | 6493 | XP_005254675.1 | |
XM_005254619.3 | 28 | 6506 | XP_005254676.2 | |
XM_005254620.3 | 28 | 6446 | XP_005254677.2 | |
XM_011521972.2 | 29 | 6707 | XP_011520274.2 | |
XM_017022521.1 | 30 | 6797 | XP_016878010.1 | |
XM_017022522.1 | 30 | 6758 | XP_016878011.1 | |
XM_017022523.1 | 30 | 6737 | XP_016878012.1 | |
XM_017022524.1 | 29 | 6557 | XP_016878013.1 | |
XM_017022525.1 | 29 | 6559 | XP_016878014.1 | |
XM_017022526.1 | 29 | 6544 | XP_016878015.1 | |
XM_017022527.1 | 29 | 6585 | XP_016878016.1 | |
XM_017022528.1 | 28 | 6469 | XP_016878017.1 | |
XM_017022529.1 | 28 | 6454 | XP_016878018.1 | |
XM_017022530.1 | 27 | 6253 | XP_016878019.1 | |
XM_017022531.1 | 30 | 6627 | XP_016878020.1 | |
AB370197.1 | 27 | 6366 | ||
AF035298.1 | 1 | 491 | ||
BX640879.1 | 28 | 6402 | CAE45936.1 | |
CR749235.1 | 20 | 5155 | CAH18091.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | ULK4P3 LOC100130111 GOLGA8T GOLGA6L7P GOLGA8M PDCD6IPP2 WHAMMP2 GOLGA8J LOC105370751 LOC101060569 LOC100289656 LOC107984752 APBA2 LOC105370746 GOLGA8R DKFZP434L187 CHRFAM7A FAM189A1 NSMCE3 TJP1 |
nsv832945 | Chr.15:29640395 - 29831518 on Build GRCh38 | Gain | LOC100130111 TJP1 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | LOC100289656 LOC100130111 LOC107984752 GOLGA6L7P PDCD6IPP2 APBA2 WHAMMP2 FAM189A1 NSMCE3 TJP1 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | ULK4P3 GOLGA6L7P PDCD6IPP2 WHAMMP2 LOC100996413 GOLGA8J LOC105370751 LOC100288637 LOC105370746 GOLGA8R DKFZP434L187 FAM189A1 LOC100130111 GOLGA8H GOLGA8T LOC101060569 LOC100289656 LOC101927579 LOC107984752 ULK4P2 ARHGAP11B APBA2 CHRFAM7A NSMCE3 TJP1 |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | ULK4P3 LOC100130111 GOLGA8T GOLGA6L7P PDCD6IPP2 WHAMMP2 GOLGA8J LOC100289656 LOC107984752 APBA2 FAM189A1 NSMCE3 TJP1 |
nsv568675 | Chr.15:29035149 - 29916451 on Build GRCh38 | Gain | LOC100130111 APBA2 FAM189A1 NSMCE3 TJP1 |
nsv832944 | Chr.15:29568858 - 29789854 on Build GRCh38 | Gain+Loss | LOC100130111 FAM189A1 TJP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |