Assay Details
Target Gene Details
Entrez Gene ID: | 9774 |
Gene Name: | BCL2 associated transcription factor 1 |
Gene Aliases: |
BTF, bK211L9.1 |
Location: |
Chr.6:136256863-136290645 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCLAF1 | NM_001077440.1 | 12 | 4873 | NP_001070908.1 |
NM_001077441.1 | 13 | 4507 | NP_001070909.1 | |
NM_001301038.1 | 13 | 5020 | NP_001287967.1 | |
NM_014739.2 | 13 | 5026 | NP_055554.1 | |
XM_005267237.3 | 12 | 4815 | XP_005267294.1 | |
XM_017011529.1 | 13 | 4494 | XP_016867018.1 | |
XM_017011530.1 | 12 | 5154 | XP_016867019.1 | |
XM_017011531.1 | 12 | 4290 | XP_016867020.1 | |
XM_017011532.1 | 14 | 5189 | XP_016867021.1 | |
BX537576.1 | 1 | 4226 | ||
BX648431.1 | 5 | 2688 | ||
BX648878.1 | 9 | 3767 | ||
D79986.1 | 13 | 5026 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |