Assay Details
Target Gene Details
Entrez Gene ID: | 79585 |
Gene Name: | coronin 7 |
Gene Aliases: |
0610011B16Rik, CRN7, POD1 |
Location: |
Chr.16:4354542-4416961 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7 | NM_001201472.1 | NP_001188401.1 | ||
NM_001201473.1 | NP_001188402.1 | |||
NM_024535.4 | NP_078811.3 | |||
AK025674.1 | BAB15211.1 | |||
AK094596.1 | ||||
AK097238.1 | ||||
AK126176.1 | ||||
AK294045.1 | ||||
AK296723.1 | ||||
AK296807.1 | ||||
BC117289.1 | ||||
BC117291.1 |
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7-PAM16 | NM_001201479.1 | NP_001188408.1 | ||
AL833954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss | CORO7-PAM16 PAM16 VASN GLIS2 GLIS2-AS1 CORO7 |
esv2659570 | Chr.16:4353172 - 4362927 on Build GRCh38 | Deletion | CORO7-PAM16 CORO7 |
esv3637696 | Chr.16:4353267 - 4362846 on Build GRCh38 | Loss | CORO7-PAM16 CORO7 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | ADCY9 HMOX2 LOC105371063 NUDT16L1 EEF2KMT SRL CDIP1 SMIM22 NAGPA-AS1 CORO7-PAM16 GLYR1 C16orf89 GLIS2-AS1 ZNF500 CORO7 C16orf71 C16orf96 SEC14L5 PPL MIR6769A PAM16 ALG1 MGRN1 VASN NAGPA NMRAL1 GLIS2 DNAJA3 UBALD1 LINC01569 RBFOX1 TFAP4 ANKS3 ROGDI UBN1 SEPT12 |
nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain | CORO7-PAM16 PAM16 VASN GLIS2 DNAJA3 GLIS2-AS1 CORO7 |
nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss | TFAP4 CORO7-PAM16 PAM16 LOC105371063 VASN GLIS2 SRL GLIS2-AS1 CORO7 LINC01569 |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain | CORO7-PAM16 PAM16 VASN NMRAL1 GLIS2 DNAJA3 CORO7 |
nsv520212 | Chr.16:4354619 - 4390827 on Build GRCh38 | Loss | CORO7-PAM16 VASN CORO7 |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain | CORO7-PAM16 HMOX2 PAM16 VASN NMRAL1 DNAJA3 CORO7 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |