Assay Details
Target Gene Details
Entrez Gene ID: | 55152 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:49015399-49021386 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DALRD3 | NM_001009996.2 | 5 | 895 | NP_001009996.1 |
NM_001276405.1 | 5 | 895 | NP_001263334.1 | |
NM_018114.5 | 5 | 912 | NP_060584.3 | |
XM_005265269.3 | 5 | 876 | XP_005265326.1 | |
XM_006713219.2 | 6 | 1031 | XP_006713282.1 | |
XM_011533891.1 | 5 | 876 | XP_011532193.1 | |
XM_011533892.1 | 5 | 1227 | XP_011532194.1 | |
XM_011533893.1 | 5 | 876 | XP_011532195.1 | |
XM_017006723.1 | 6 | 1031 | XP_016862212.1 | |
AK001358.1 | 4 | 880 | BAA91647.1 | |
AK093054.1 | 2 | 1174 | ||
AK093204.1 | 5 | 876 | BAC04095.1 | |
AK093294.1 | 4 | 683 | BAC04123.1 | |
AK301352.1 | 5 | 863 | ||
AK308273.1 | 6 | 989 | ||
BC032440.1 | 5 | 858 | AAH32440.2 | |
BC047683.1 | 5 | 857 | AAH47683.1 | |
BC054493.1 | 5 | 851 | AAH54493.1 | |
BX648719.1 | 3 | 542 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954488 | Chr.3:49002068 - 49019667 on Build GRCh38 | Deletion | DALRD3 P4HTM WDR6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |