Assay Details
Target Gene Details
Entrez Gene ID: | 1499 |
Gene Name: | catenin beta 1 |
Gene Aliases: |
CTNNB, MRD19, armadillo |
Location: |
Chr.3:41199451-41240448 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTNNB1 | NM_001098209.1 | 3 | 443 | NP_001091679.1 |
NM_001098210.1 | 3 | 443 | NP_001091680.1 | |
NM_001904.3 | 3 | 443 | NP_001895.1 | |
XM_005264886.2 | 3 | 457 | XP_005264943.1 | |
XM_006712983.1 | 3 | 355 | XP_006713046.1 | |
XM_006712984.1 | 3 | 355 | XP_006713047.1 | |
XM_006712985.1 | 3 | 417 | XP_006713048.1 | |
XM_017005738.1 | 3 | 457 | XP_016861227.1 | |
AB451264.1 | 1 | 175 | ||
AB451392.1 | 1 | 175 | ||
AK289932.1 | 4 | 331 | ||
AK294778.1 | 4 | 556 | ||
AK296790.1 | 3 | 389 | ||
AK299954.1 | ||||
AK314776.1 | 3 | 389 | ||
BC058926.1 | 3 | 385 | AAH58926.1 | |
DA216720.1 | 3 | 443 | ||
X87838.1 | 3 | 389 | CAA61107.1 | |
Z19054.1 | 3 | 375 | CAA79497.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |