Assay Details
Target Gene Details
Entrez Gene ID: | 9787 |
Gene Name: | DLG associated protein 5 |
Gene Aliases: |
DLG7, HURP |
Location: |
Chr.14:55148116-55191678 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP5 | NM_001146015.1 | 17 | 2513 | NP_001139487.1 |
NM_014750.4 | 17 | 2513 | NP_055565.3 | |
XM_017021840.1 | 17 | 2405 | XP_016877329.1 | |
AB076695.1 | 17 | 2513 | BAB97376.1 | |
AI174844.1 | 2 | 205 | ||
AK291847.1 | 17 | 2390 | ||
AK297480.1 | 16 | 2266 | ||
AK299338.1 | 17 | 2390 | ||
BC010658.1 | 17 | 2405 | AAH10658.2 | |
BC016276.2 | 16 | 2299 | AAH16276.2 | |
BT007344.1 | 15 | 2053 | AAP36008.1 | |
BX248255.1 | 17 | 2379 | CAD62583.1 | |
D13633.3 | 17 | 2386 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |