Assay Details
Target Gene Details
Entrez Gene ID: | 10549 |
Gene Name: | peroxiredoxin 4 |
Gene Aliases: |
AOE37-2, AOE372, HEL-S-97n, PRX-4 |
Location: |
Chr.X:23664260-23686399 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRDX4 | NM_006406.1 | 2 | 341 | NP_006397.1 |
XM_005274438.1 | 2 | 516 | XP_005274495.1 | |
XM_017029230.1 | 2 | 318 | XP_016884719.1 | |
XM_017029231.1 | 2 | 516 | XP_016884720.1 | |
BC003609.2 | 2 | 301 | AAH03609.1 | |
BC007107.1 | 2 | 345 | AAH07107.1 | |
BC016770.1 | 2 | 355 | AAH16770.1 | |
CR541668.1 | 2 | 298 | CAG46469.1 | |
CR541705.1 | 2 | 298 | CAG46506.1 | |
FJ224297.1 | 2 | 341 | ||
U25182.1 | 2 | 341 | AAB95175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526897 | Chr.X:23668070 - 23683545 on Build GRCh38 | Gain | PRDX4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |