Assay Details
Target Gene Details
Entrez Gene ID: | 27158 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
NR1, bA350O14.9 |
Location: |
Chr.9:137205667-137219361 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDOR1 | NM_001144026.2 | 14 | 1962 | NP_001137498.1 |
NM_001144027.2 | 13 | 1811 | NP_001137499.1 | |
NM_001144028.2 | 14 | 1914 | NP_001137500.1 | |
NM_014434.3 | 14 | 1935 | NP_055249.1 | |
XM_011518545.1 | 13 | 1850 | XP_011516847.1 | |
XM_011518547.1 | 14 | 1752 | XP_011516849.1 | |
XM_011518548.2 | 13 | 3152 | XP_011516850.1 | |
XM_017014626.1 | XP_016870115.1 | |||
XM_017014627.1 | 14 | 1913 | XP_016870116.1 | |
AF199509.1 | 14 | 1871 | AAF25205.1 | |
AK026089.1 | 14 | 1934 | ||
AK074403.1 | 13 | 1772 | ||
AK225995.1 | 14 | 1933 | ||
AK290026.1 | ||||
AY077845.1 | AAL77754.1 | |||
BC015735.1 | 14 | 1871 | AAH15735.1 | |
BC093782.1 | AAH93782.1 | |||
BC111943.1 | AAI11944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13010n54 | Chr.9:137191539 - 137233413 on Build GRCh38 | Loss | TPRN RNF224 TMEM203 NDOR1 CYSRT1 SLC34A3 RNF208 |
nsv616012 | Chr.9:137197680 - 137223834 on Build GRCh38 | Loss | TPRN TMEM203 NDOR1 RNF208 |
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | LOC101930307 SSNA1 RNF224 TMEM203 TMEM210 NDOR1 LRRC26 MAN1B1-AS1 SLC34A3 DPP7 MIR3621 TPRN CYSRT1 GRIN1 ANAPC2 RNF208 UAP1L1 MAN1B1 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | STPG3 SSNA1 RNF224 TMEM203 EXD3 TUBB4B NDOR1 TOR4A SLC34A3 C9orf173-AS1 FAM166A TPRN NELFB CYSRT1 NRARP ANAPC2 RNF208 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | LOC101930307 RNF224 TMEM203 C9orf172 LCN12 EXD3 CLIC3 PNPLA7 LRRC26 MIR4292 C9orf173-AS1 FAM166A ENTPD2 CYSRT1 MIR4479 MIR7114 GRIN1 SAPCD2 ANAPC2 NSMF RNF208 MAN1B1-AS1 DPP7 MIR3621 CCDC183-AS1 NRARP FUT7 C9orf142 NPDC1 MAN1B1 SSNA1 LCNL1 EDF1 C9orf139 TOR4A PRR31 NELFB TRAF2 PHPT1 NOXA1 ABCA2 STPG3 TMEM210 TUBB4B CCDC183 NDOR1 SLC34A3 RABL6 MAMDC4 TPRN FBXW5 C8G PTGDS ENTPD8 UAP1L1 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | STPG3 SSNA1 RNF224 TMEM203 EXD3 TMEM210 TUBB4B NDOR1 TOR4A LRRC26 SLC34A3 C9orf173-AS1 MIR3621 FAM166A TPRN NELFB CYSRT1 NRARP GRIN1 ANAPC2 RNF208 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | STPG3 SSNA1 RNF224 TMEM203 EXD3 TMEM210 TUBB4B NDOR1 TOR4A LRRC26 SLC34A3 C9orf173-AS1 MIR3621 FAM166A TPRN NELFB CYSRT1 NRARP ENTPD8 GRIN1 ANAPC2 RNF208 NOXA1 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | LOC101930307 RNF224 TMEM203 C9orf172 LCN12 EXD3 CLIC3 LCN10 LRRC26 MIR4292 LCN6 C9orf173-AS1 FAM166A LCN15 MIR126 SNHG7 ENTPD2 CYSRT1 MIR4479 GRIN1 SAPCD2 ANAPC2 RNF208 AGPAT2 MAN1B1-AS1 DPP7 HSPC324 MIR3621 TMEM141 CCDC183-AS1 SNORA17A NRARP FUT7 C9orf142 NPDC1 LOC100128593 FAM69B MAN1B1 SSNA1 LCNL1 EDF1 C9orf139 TOR4A PRR31 NELFB MIR6722 SNORA17B LCN8 TRAF2 PHPT1 ABCA2 STPG3 TMEM210 TUBB4B CCDC183 NDOR1 SLC34A3 RABL6 MAMDC4 TPRN FBXW5 C8G LINC01451 PTGDS EGFL7 UAP1L1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |