Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENTPD8 | NM_001033113.1 | NP_001028285.1 | ||
NM_198585.2 | NP_940987.2 | |||
XM_011518665.2 | XP_011516967.1 | |||
XM_011518666.2 | XP_011516968.1 | |||
XM_011518668.1 | XP_011516970.1 | |||
AY359088.1 | AAQ89446.1 | |||
AY430414.1 | AAR04374.1 | |||
AY903953.1 | 3 | 846 | AAW83515.1 | |
AY903954.1 | AAW83516.1 | |||
BC141810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss | NSMF MIR7114 ENTPD8 |
nsv1049808 | Chr.9:137356366 - 137438026 on Build GRCh38 | Gain | NOXA1 EXD3 ENTPD8 |
nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion | PNPLA7 NSMF NOXA1 EXD3 MIR7114 ENTPD8 |
nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion | NOXA1 EXD3 ENTPD8 |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss | NOXA1 EXD3 ENTPD8 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | SAPCD2 NPDC1 MAN1B1 ANAPC2 RNF208 CLIC3 SLC34A3 TMEM203 NRARP TUBB4B PHPT1 RABL6 LCN12 MIR7114 TRAF2 TOR4A LOC101930307 DPP7 RNF224 C8G TMEM210 EXD3 STPG3 MAN1B1-AS1 C9orf142 C9orf139 EDF1 TPRN SSNA1 MAMDC4 LRRC26 C9orf173-AS1 FAM166A GRIN1 CYSRT1 PTGDS CCDC183 NOXA1 NDOR1 MIR4479 CCDC183-AS1 MIR4292 LCNL1 PRR31 FBXW5 ENTPD2 ABCA2 PNPLA7 NSMF FUT7 NELFB MIR3621 UAP1L1 ENTPD8 C9orf172 |
esv3622034 | Chr.9:137433077 - 137439026 on Build GRCh38 | Gain | NOXA1 ENTPD8 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | NOXA1 TPRN NDOR1 SSNA1 LRRC26 TOR4A ANAPC2 RNF224 RNF208 C9orf173-AS1 FAM166A GRIN1 SLC34A3 TMEM203 TMEM210 NRARP EXD3 NELFB TUBB4B CYSRT1 MIR3621 STPG3 ENTPD8 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |