Assay Details
Target Gene Details
Entrez Gene ID: | 113178 |
Gene Name: | secretory carrier membrane protein 4 |
Gene Aliases: |
SCAMP-4 |
Location: |
Chr.19:1905374-1926013 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCAMP4 | NM_079834.2 | 6 | 551 | NP_524558.1 |
AK074967.1 | 6 | 526 | BAC11322.1 | |
AK091166.1 | 1 | 986 | ||
AK092056.1 | 5 | 419 | BAC03797.1 | |
AK307641.1 | 5 | 477 | ||
AK307700.1 | 5 | 475 | ||
AL832731.1 | 4 | 380 | ||
BC011747.2 | 6 | 513 | AAH11747.1 | |
BC016509.1 | 6 | 525 | AAH16509.1 | |
BC062598.1 | 6 | 524 | AAH62598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss | CSNK1G2-AS1 ADAT3 ATP8B3 IZUMO4 MKNK2 REXO1 KLF16 BTBD2 ABHD17A AP3D1 ONECUT3 MIR1909 MOB3A SCAMP4 LOC100288123 CSNK1G2 |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | TCF3 MBD3 BTBD2 MIR1909 RPS15 SCAMP4 REEP6 LOC100288123 CSNK1G2 GAMT CSNK1G2-AS1 ADAT3 UQCR11 ATP8B3 REXO1 KLF16 NDUFS7 PCSK4 C19orf25 ABHD17A DAZAP1 MUM1 PLK5 ONECUT3 APC2 MEX3D ADAMTSL5 |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss | CSNK1G2-AS1 ADAT3 ATP8B3 IZUMO4 MKNK2 REXO1 KLF16 BTBD2 ABHD17A MIR1909 MOB3A SCAMP4 LOC100288123 CSNK1G2 |
esv2571215 | Chr.19:1922944 - 1923921 on Build GRCh38 | Insertion | SCAMP4 |
esv3643413 | Chr.19:1922441 - 1923828 on Build GRCh38 | Loss | SCAMP4 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | TCF3 MBD3 MKNK2 BTBD2 MIR1909 RPS15 SCAMP4 REEP6 LOC100288123 CSNK1G2 GAMT CSNK1G2-AS1 ADAT3 UQCR11 ATP8B3 REXO1 KLF16 NDUFS7 PCSK4 C19orf25 ABHD17A DAZAP1 MUM1 PLK5 ONECUT3 APC2 MEX3D ADAMTSL5 |
nsv1160561 | Chr.19:1884553 - 2003982 on Build GRCh38 | Deletion | CSNK1G2-AS1 ADAT3 SCAMP4 BTBD2 CSNK1G2 ABHD17A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |