Assay Details
Target Gene Details
Entrez Gene ID: | 29070 |
Gene Name: | coiled-coil domain containing 113 |
Gene Aliases: |
HSPC065 |
Location: |
Chr.16:58196863-58283836 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC113 | NM_001142302.1 | 8 | 1300 | NP_001135774.1 |
NM_014157.3 | 9 | 1462 | NP_054876.2 | |
XM_011523038.2 | 9 | 1317 | XP_011521340.1 | |
XM_011523039.2 | 11 | 2210 | XP_011521341.1 | |
XM_011523040.2 | 9 | 1735 | XP_011521342.1 | |
XM_011523041.2 | 9 | 1303 | XP_011521343.1 | |
XM_017023176.1 | 10 | 1625 | XP_016878665.1 | |
XM_017023177.1 | 8 | 1319 | XP_016878666.1 | |
AK299066.1 | ||||
AL136785.1 | 9 | 1439 | CAB66719.1 | |
BC126445.1 | 9 | 1431 | ||
BC126447.1 | 9 | 1431 |
Target Gene Details
Entrez Gene ID: | 221191 |
Gene Name: | protease, serine 54 |
Gene Aliases: |
CT67, KLKBL4 |
Location: |
Chr.16:58279997-58295047 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS54 | NM_001080492.1 | NP_001073961.1 | ||
NM_001305173.1 | NP_001292102.1 | |||
NM_001305174.1 | NP_001292103.1 | |||
AK302175.1 | ||||
AL137484.1 | CAB70765.1 | |||
BC057843.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |