Assay Details
Target Gene Details
Entrez Gene ID: | 81033 |
Gene Name: | potassium voltage-gated channel subfamily H member 6 |
Gene Aliases: |
ERG-2, ERG2, HERG2, Kv11.2, hERG-2 |
Location: |
Chr.17:63523334-63548977 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNH6 | NM_001278919.1 | 4 | 671 | NP_001265848.1 |
NM_001278920.1 | 4 | 538 | NP_001265849.1 | |
NM_030779.3 | 4 | 671 | NP_110406.1 | |
NM_173092.2 | 4 | 671 | NP_775115.1 | |
XM_011525308.2 | 4 | 671 | XP_011523610.1 | |
XM_011525309.2 | 4 | 671 | XP_011523611.1 | |
XM_011525310.2 | 4 | 671 | XP_011523612.1 | |
XM_011525311.1 | 4 | 671 | XP_011523613.1 | |
XM_011525312.1 | 4 | 671 | XP_011523614.1 | |
XM_011525313.1 | 4 | 671 | XP_011523615.1 | |
XM_017025175.1 | 4 | 671 | XP_016880664.1 | |
XM_017025176.1 | 4 | 671 | XP_016880665.1 | |
XM_017025177.1 | 4 | 671 | XP_016880666.1 | |
XM_017025178.1 | 4 | 671 | XP_016880667.1 | |
XM_017025179.1 | 4 | 671 | XP_016880668.1 | |
XM_017025180.1 | 4 | 507 | XP_016880669.1 | |
AF311913.1 | 4 | 602 | AAG40871.1 | |
AK091877.1 | 4 | 671 | BAC03764.1 | |
AK296498.1 | 4 | 644 | ||
AK298363.1 | 4 | 513 | ||
BC006334.2 | 4 | 649 | AAH06334.1 | |
CD632730.1 | 2 | 497 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3640998 | Chr.17:63526371 - 63539713 on Build GRCh38 | Loss | KCNH6 |
nsv952355 | Chr.17:63522640 - 63550539 on Build GRCh38 | Deletion | KCNH6 DCAF7 |
esv2675657 | Chr.17:63526487 - 63539627 on Build GRCh38 | Deletion | KCNH6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |