Assay Details
Target Gene Details
Entrez Gene ID: | 100 |
Gene Name: | adenosine deaminase |
Gene Aliases: |
- |
Location: |
Chr.20:44619519-44651758 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADA | NM_000022.3 | NP_000013.2 | ||
NM_001322050.1 | NP_001308979.1 | |||
NM_001322051.1 | NP_001308980.1 | |||
NR_136160.1 | ||||
AK223397.1 | BAD97117.1 | |||
BC007678.2 | AAH07678.1 | |||
BC040226.1 | AAH40226.1 | |||
BI601734.1 | ||||
CD014053.1 | ||||
CD014056.1 | ||||
HY102642.1 | ||||
KJ534760.1 | ||||
KU177892.1 | ||||
KU177893.1 | ||||
KU177894.1 | ||||
KU177895.1 | ||||
X02994.1 | CAA26734.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |