Assay Details
Target Gene Details
Entrez Gene ID: | 83875 |
Gene Name: | beta-carotene oxygenase 2 |
Gene Aliases: |
B-DIOX-II, BCDO2 |
Location: |
Chr.11:112175485-112218929 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCO2 | NM_001037290.2 | 7 | 919 | NP_001032367.2 |
NM_001256397.1 | 7 | 919 | NP_001243326.1 | |
NM_001256398.1 | 5 | 812 | NP_001243327.1 | |
NM_001256400.1 | 6 | 714 | NP_001243329.1 | |
NM_031938.5 | 7 | 1031 | NP_114144.4 | |
AB096252.1 | 7 | 885 | ||
AF276432.1 | 7 | 1528 | AAK69433.1 | |
AJ290393.1 | 7 | 881 | CAC27994.1 | |
AK027801.1 | 7 | 914 | BAB55379.1 | |
AK091783.1 | 5 | 947 | BAC03747.1 | |
AK297853.1 | 6 | 705 | ||
AK315300.1 | 7 | 1023 | ||
BC041656.1 | 5 | 775 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761708 | Chr.11:112176838 - 112212857 on Build GRCh38 | Loss |
![]() |
esv3579908 | Chr.11:112173319 - 112217076 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)