Assay Details
Target Gene Details
Entrez Gene ID: | 114786 |
Gene Name: | XK related 4 |
Gene Aliases: |
XRG4 |
Location: |
Chr.8:55102457-55526155 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XKR4 | NM_052898.1 | 1 | 618 | NP_443130.1 |
AB067476.1 | 1 | 152 | BAB67782.1 | |
AY534241.1 | 1 | 618 | AAT07090.1 |