Assay Details
Target Gene Details
Entrez Gene ID: | 8405 |
Gene Name: | speckle type BTB/POZ protein |
Gene Aliases: |
BTBD32, TEF2 |
Location: |
Chr.17:49598884-49678163 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPOP | NM_001007226.1 | 8 | 1050 | NP_001007227.1 |
NM_001007227.1 | 7 | 928 | NP_001007228.1 | |
NM_001007228.1 | 6 | 876 | NP_001007229.1 | |
NM_001007229.1 | 7 | 927 | NP_001007230.1 | |
NM_001007230.1 | 8 | 979 | NP_001007231.1 | |
NM_003563.3 | 7 | 998 | NP_003554.1 | |
XM_005257723.4 | 7 | 1222 | XP_005257780.1 | |
XM_005257724.4 | 6 | 1152 | XP_005257781.1 | |
XM_017025204.1 | 5 | 665 | XP_016880693.1 | |
AJ000644.1 | 6 | 736 | CAA04199.1 | |
AK222589.1 | 6 | 701 | BAD96309.1 | |
AK293045.1 | 7 | 1209 | ||
AK293587.1 | 6 | 682 | ||
AK300616.1 | 6 | 700 | ||
AK303268.1 | 5 | 622 | ||
AK312691.1 | 6 | 712 | ||
AK316366.1 | 8 | 777 | ||
BC001269.1 | 7 | 882 | AAH01269.1 | |
BC003385.1 | 6 | 693 | AAH03385.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |