Assay Details
Target Gene Details
Entrez Gene ID: | 493860 |
Gene Name: | coiled-coil domain containing 73 |
Gene Aliases: |
NY-SAR-79 |
Location: |
Chr.11:32602080-32794658 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC73 | NM_001008391.3 | 18 | 3880 | NP_001008392.2 |
XM_011520139.2 | XP_011518441.1 | |||
AA521271.1 | 1 | 244 | ||
AK128159.1 | BAC87304.1 | |||
AK131470.1 | BAD18615.1 | |||
AW088759.1 | 1 | 235 |
Target Gene Details
Entrez Gene ID: | 10480 |
Gene Name: | eukaryotic translation initiation factor 3 subunit M |
Gene Aliases: |
B5, GA17, PCID1, TANGO7, hfl-B5 |
Location: |
Chr.11:32583767-32602873 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF3M | NM_001307929.1 | NP_001294858.1 | ||
NM_006360.5 | NP_006351.2 | |||
XM_011519840.2 | XP_011518142.1 | |||
AF064603.1 | AAC17108.1 | |||
AF277183.1 | AAK07542.1 | |||
AK131064.1 | ||||
AK222564.1 | BAD96284.1 | |||
AK292139.1 | ||||
AK304378.1 | ||||
AK312512.1 | ||||
AV713889.1 | ||||
AY769947.1 | AAX12524.1 | |||
BC019103.2 | AAH19103.1 | |||
BC051292.1 | AAH51292.1 | |||
CK002866.1 | ||||
CR450300.1 | CAG29296.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |