Assay Details
Target Gene Details
Entrez Gene ID: | 5133 |
Gene Name: | programmed cell death 1 |
Gene Aliases: |
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1 |
Location: |
Chr.2:241849881-241858908 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDCD1 | NM_005018.2 | 5 | 817 | NP_005009.2 |
XM_017004293.1 | 5 | 816 | XP_016859782.1 | |
AK313848.1 | 5 | 819 | ||
AY206416.1 | AAO25116.1 | |||
AY238517.1 | 5 | 749 | AAO63583.1 | |
BC074740.2 | 5 | 795 | AAH74740.1 | |
KJ865859.1 | 5 | 749 | ||
KJ865860.1 | 5 | 749 | ||
KJ865861.1 | 5 | 749 | ||
U64863.1 | 5 | 817 | AAC51773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv955302 | Chr.2:241840449 - 241891349 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv998059 | Chr.2:241830506 - 242001132 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3893582 | Chr.2:241821357 - 241977794 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv997984 | Chr.2:241723206 - 242001132 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv165e55 | Chr.2:241576690 - 242072479 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv584991 | Chr.2:241805306 - 242006956 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4201n100 | Chr.2:241793613 - 242017881 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1002941 | Chr.2:241703438 - 242147293 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470533 | Chr.2:241816852 - 242101996 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4200n100 | Chr.2:241786337 - 242092523 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)