Assay Details
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001128325.2 | 4 | 529 | NP_001121797.1 |
NM_001199021.1 | 5 | 883 | NP_001185950.1 | |
NM_012445.3 | 3 | 587 | NP_036577.1 | |
AB027466.1 | 3 | 533 | BAA85892.1 | |
AK074618.1 | 3 | 394 | BAC11092.1 | |
AK074770.1 | 3 | 525 | BAC11196.1 | |
AK130164.1 | 3 | 305 | ||
AY358948.1 | 3 | 532 | AAQ89307.1 | |
BC002707.2 | 3 | 317 | AAH02707.1 | |
BC036341.1 | 3 | 526 | AAH36341.1 | |
BY797606.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | LOC101928521 LOC100130872 MFSD7 NKX1-1 LOC107986211 TMED11P CTBP1 MAEA MYL5 ATP5I LOC101928548 SLC26A1 IDUA RNF212 UVSSA DGKQ PCGF3 GAK PDE6B LOC100129917 CTBP1-AS FGFRL1 TMEM175 CPLX1 CRIPAK LOC105374344 SPON2 CTBP1-AS2 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | CTBP1-AS MAEA LOC100130872 NKX1-1 UVSSA LOC101928548 CRIPAK SPON2 CTBP1 CTBP1-AS2 |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | CTBP1-AS LOC100130872 LOC101928548 SPON2 CTBP1 |
nsv519106 | Chr.4:1165203 - 1181073 on Build GRCh38 | Loss | SPON2 |
nsv1012892 | Chr.4:1093083 - 1182842 on Build GRCh38 | Loss | RNF212 TMED11P LOC105374344 SPON2 |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | CTBP1-AS RNF212 MAEA LOC100130872 LOC101928548 TMED11P LOC105374344 SPON2 CTBP1 CTBP1-AS2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |