Assay Details
Target Gene Details
Entrez Gene ID: | 6657 |
Gene Name: | SRY-box 2 |
Gene Aliases: |
ANOP3, MCOPS3 |
Location: |
Chr.3:181711924-181714436 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOX2 | NM_003106.3 | 1 | 1485 | NP_003097.1 |
AW016610.1 | 1 | 364 | ||
BC013923.2 | AAH13923.1 | |||
L07335.1 | AAA35997.1 | |||
Z31560.1 | CAA83435.1 |
Target Gene Details
Entrez Gene ID: | 347689 |
Gene Name: | SOX2 overlapping transcript |
Gene Aliases: |
NCRNA00043, SOX2OT |
Location: |
Chr.3:181056680-181742228 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOX2-OT | NR_004053.3 | |||
NR_075089.1 | ||||
NR_075090.1 | ||||
NR_075091.1 | ||||
NR_075092.1 | ||||
NR_075093.1 | ||||
AK022826.1 | ||||
AL157425.1 | ||||
DA310380.1 | ||||
DA366949.1 | ||||
JN711430.1 | ||||
JN882275.1 | ||||
JQ408703.1 | ||||
JX110620.1 | ||||
JX504683.1 | ||||
KF981435.1 | ||||
KJ415055.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |