Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AI796350.1 | ||||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | 11 | 1913 | BAC04000.1 | |
AK292004.1 | ||||
AK300915.1 | 11 | 1934 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | EDC4 LOC100505942 ELMO3 SLC12A4 PARD6A ENKD1 RANBP10 AGRP KCTD19 PSKH1 DDX28 FBXL8 DPEP2 THAP11 CARMIL2 FAM65A E2F4 GFOD2 C16orf86 NUTF2 ATP6V0D1 LOC105369155 LOC107984813 DUS2 HSD11B2 MIR328 TMEM208 LCAT CTCF LRRC29 HSF4 KIAA0895L ACD PLEKHG4 CTRL NRN1L TPPP3 DPEP3 TSNAXIP1 FHOD1 PSMB10 LOC100131303 LRRC36 CENPT SLC9A5 TRADD ZDHHC1 NOL3 EXOC3L1 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | MIR328 TMEM208 FBXL8 LRRC29 HSF4 ELMO3 KIAA0895L C16orf70 PLEKHG4 E2F4 FHOD1 SLC9A5 B3GNT9 TRADD NOL3 LOC105369155 EXOC3L1 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | MIR328 TMEM208 FBXL8 LRRC29 HSF4 ELMO3 KIAA0895L C16orf70 E2F4 FHOD1 B3GNT9 TRADD NOL3 LOC105369155 EXOC3L1 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | MIR328 FBXL8 E2F4 LRRC29 HSF4 ELMO3 KIAA0895L NOL3 LOC105369155 EXOC3L1 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | MIR328 TMEM208 E2F4 FHOD1 LRRC29 SLC9A5 ELMO3 KIAA0895L PLEKHG4 LOC105369155 EXOC3L1 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | MIR328 TMEM208 LRRC29 ELMO3 KIAA0895L PLEKHG4 E2F4 FHOD1 SLC9A5 KCTD19 NOL3 LOC105369155 EXOC3L1 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | EDC4 LOC100505942 ELMO3 SLC12A4 PARD6A ENKD1 RANBP10 AGRP KCTD19 B3GNT9 PSKH1 DDX28 FBXL8 DPEP2 C16orf70 THAP11 CARMIL2 FAM65A E2F4 GFOD2 C16orf86 NUTF2 ATP6V0D1 LOC105369155 LOC107984813 DUS2 HSD11B2 MIR328 TMEM208 LCAT CTCF LRRC29 HSF4 KIAA0895L ACD PLEKHG4 CTRL NRN1L TPPP3 DPEP3 TSNAXIP1 FHOD1 PSMB10 LOC100131303 LRRC36 CENPT SLC9A5 TRADD ZDHHC1 NOL3 EXOC3L1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |