Assay Details
Target Gene Details
Entrez Gene ID: | 4793 |
Gene Name: | NFKB inhibitor beta |
Gene Aliases: |
IKBB, TRIP9 |
Location: |
Chr.19:38899503-38908894 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFKBIB | NM_001243116.1 | NP_001230045.1 | ||
NM_002503.4 | NP_002494.2 | |||
NR_040515.1 | ||||
XM_006723226.2 | XP_006723289.2 | |||
XM_006723227.3 | XP_006723290.3 | |||
AB209784.1 | BAD93021.1 | |||
AB362577.1 | ||||
AK057862.1 | BAB71599.1 | |||
AK290569.1 | ||||
BC015528.1 | AAH15528.1 | |||
BI520169.1 | ||||
BM008311.1 | ||||
BM449613.1 | ||||
BQ440907.1 | ||||
BT006743.1 | AAP35389.1 | |||
DA633677.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |