Assay Details
Target Gene Details
Entrez Gene ID: | 29126 |
Gene Name: | CD274 molecule |
Gene Aliases: |
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 |
Location: |
Chr.9:5450503-5470567 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD274 | NM_001267706.1 | NP_001254635.1 | ||
NM_014143.3 | NP_054862.1 | |||
NR_052005.1 | ||||
AF177937.1 | AAF25807.1 | |||
AF233516.1 | AAG18508.1 | |||
AK001894.1 | BAA91966.1 | |||
AK314567.1 | ||||
AY254342.1 | AAP13470.1 | |||
AY291313.1 | AAP42144.1 | |||
AY714881.1 | AAU09634.1 | |||
BC069381.1 | AAH69381.1 | |||
BC074984.2 | AAH74984.1 | |||
BC113734.1 | ||||
BC113736.1 | ||||
DQ286582.1 | ABB90152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv613196 | Chr.9:5345673 - 5514372 on Build GRCh38 | Gain | CD274 LOC107987017 PLGRKT PDCD1LG2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |