Assay Details
Target Gene Details
Entrez Gene ID: | 111 |
Gene Name: | adenylate cyclase 5 |
Gene Aliases: |
AC5, FDFM |
Location: |
Chr.3:123282296-123448632 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY5 | NM_001199642.1 | 21 | 2872 | NP_001186571.1 |
NM_183357.2 | 21 | 3847 | NP_899200.1 | |
XM_005247077.3 | 22 | 4009 | XP_005247134.1 | |
XM_005247078.1 | 22 | 2947 | XP_005247135.1 | |
XM_006713483.1 | 22 | 2943 | XP_006713546.1 | |
XM_006713484.1 | 22 | 2948 | XP_006713547.1 | |
XM_011512359.2 | 22 | 3807 | XP_011510661.1 | |
XM_011512360.2 | 22 | 3004 | XP_011510662.1 | |
XM_011512361.1 | 22 | 2911 | XP_011510663.1 | |
XM_017005638.1 | 22 | 2839 | XP_016861127.1 | |
XM_017005639.1 | 22 | 2839 | XP_016861128.1 | |
XM_017005640.1 | 22 | 2935 | XP_016861129.1 | |
AF497517.1 | AAM94374.1 | |||
AK093840.1 | 17 | 1951 | ||
AK124691.1 | 23 | 3359 | ||
AK294565.1 | ||||
AK302711.1 | 9 | 1418 | ||
AK316488.1 | ||||
BX647739.1 | 18 | 2349 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3976 | Chr.3:123251695 - 123286279 on Build GRCh38 | Insertion | SEC22A ADCY5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |