Assay Details
Target Gene Details
Entrez Gene ID: | 128954 |
Gene Name: | GRB2 associated binding protein family member 4 |
Gene Aliases: |
- |
Location: |
Chr.22:16961936-17008281 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAB4 | NM_001037814.1 | 10 | 1910 | NP_001032903.1 |
XM_011546114.2 | 8 | 1506 | XP_011544416.1 | |
XM_011546115.1 | 9 | 1447 | XP_011544417.1 | |
XM_011546117.1 | 8 | 1203 | XP_011544419.1 | |
XM_017028575.1 | 10 | 1965 | XP_016884064.1 | |
XM_017028576.1 | 8 | 1134 | XP_016884065.1 | |
AB076978.1 | 10 | 1910 | BAD06454.1 | |
AK057252.1 | 9 | 2502 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2763696 | Chr.22:16384236 - 16983056 on Build GRCh38 | Gain | GAB4 XKR3 ANKRD62P1-PARP4P3 CCT8L2 ACTR3BP6 LOC101929350 HSFY1P1 TPTEP1 |
esv3647204 | Chr.22:16939332 - 17123705 on Build GRCh38 | Gain | GAB4 CECR7 IL17RA LOC100996342 CECR6 |
dgv4460n100 | Chr.22:16384224 - 17135620 on Build GRCh38 | Gain | GAB4 XKR3 ANKRD62P1-PARP4P3 CCT8L2 CECR7 ACTR3BP6 IL17RA LOC101929350 HSFY1P1 LOC100996342 CECR6 TPTEP1 |
nsv588077 | Chr.22:16674094 - 16972076 on Build GRCh38 | Gain | GAB4 XKR3 ANKRD62P1-PARP4P3 LOC101929350 HSFY1P1 |
dgv4463n100 | Chr.22:16749141 - 17081023 on Build GRCh38 | Gain | GAB4 XKR3 CECR7 HSFY1P1 |
nsv527637 | Chr.22:16437596 - 16981295 on Build GRCh38 | Gain | GAB4 XKR3 ANKRD62P1-PARP4P3 CCT8L2 ACTR3BP6 LOC101929350 HSFY1P1 TPTEP1 |
esv3575406 | Chr.22:16408174 - 16982154 on Build GRCh38 | Gain | GAB4 XKR3 ANKRD62P1-PARP4P3 CCT8L2 ACTR3BP6 LOC101929350 HSFY1P1 TPTEP1 |
nsv1064641 | Chr.22:16757380 - 17233805 on Build GRCh38 | Gain | GAB4 XKR3 CECR7 CECR5 IL17RA HSFY1P1 CECR5-AS1 CECR1 LOC100996342 CECR6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |