Assay Details
Target Gene Details
Entrez Gene ID: | 84899 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 4 |
Gene Aliases: |
- |
Location: |
Chr.13:100603836-100675025 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMTC4 | NM_001079669.2 | 18 | 2427 | NP_001073137.1 |
NM_001286453.1 | 16 | 2094 | NP_001273382.1 | |
NM_032813.3 | 19 | 2637 | NP_116202.2 | |
XM_011521121.2 | 20 | 2789 | XP_011519423.1 | |
XM_011521122.2 | 20 | 2705 | XP_011519424.1 | |
XM_011521123.2 | 20 | 2866 | XP_011519425.1 | |
XM_011521124.2 | 19 | 2494 | XP_011519426.1 | |
XM_017020796.1 | 19 | 2692 | XP_016876285.1 | |
XM_017020797.1 | 18 | 2431 | XP_016876286.1 | |
XM_017020798.1 | 18 | 2527 | XP_016876287.1 | |
XM_017020799.1 | 17 | 2212 | XP_016876288.1 | |
AF339792.1 | 16 | 2009 | ||
AK027530.1 | 13 | 1520 | BAB55179.1 | |
AK056409.1 | 19 | 2496 | ||
AK124599.1 | 1 | 3098 | ||
AK290345.1 | 18 | 2287 | ||
AK299859.1 | 16 | 1933 | ||
BX647956.1 | 18 | 2318 | ||
BX648008.1 | 18 | 2271 | ||
DA368450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1037414 | Chr.13:100521000 - 100622872 on Build GRCh38 | Loss | TMTC4 GGACT PCCA |