Assay Details
Target Gene Details
Entrez Gene ID: | 84262 |
Gene Name: | proteasome assembly chaperone 3 |
Gene Aliases: |
C7orf48, PAC3 |
Location: |
Chr.7:1567332-1570032 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMG3 | NM_001134340.1 | 3 | 691 | NP_001127812.1 |
NM_032302.3 | 2 | 1300 | NP_115678.1 | |
BC004308.1 | 2 | 879 | AAH04308.1 | |
BC027171.1 | 2 | 1301 | AAH27171.1 | |
CN261807.1 | 2 | 685 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950786 | Chr.7:1381265 - 1675664 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2759499 | Chr.7:1159459 - 1650311 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv830879 | Chr.7:1493586 - 1650311 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3611812 | Chr.7:1557046 - 1567824 on Build GRCh38 | Gain |
![]() ![]() |
nsv5615 | Chr.7:1528557 - 1573208 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
nsv605820 | Chr.7:1518331 - 1574625 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)