Assay Details
Target Gene Details
Entrez Gene ID: | 10525 |
Gene Name: | hypoxia up-regulated 1 |
Gene Aliases: |
GRP-170, Grp170, HSP12A, ORP-150, ORP150 |
Location: |
Chr.11:119044185-119057246 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYOU1 | NM_001130991.2 | 13 | 1537 | NP_001124463.1 |
NM_006389.4 | 13 | 1670 | NP_006380.1 | |
XM_005271392.3 | 13 | 1531 | XP_005271449.1 | |
XM_005271393.3 | 13 | 1511 | XP_005271450.1 | |
XM_005271394.3 | 13 | 1578 | XP_005271451.1 | |
XM_017017095.1 | 13 | 1566 | XP_016872584.1 | |
XM_017017096.1 | 13 | 1587 | XP_016872585.1 | |
XM_017017097.1 | 13 | 1510 | XP_016872586.1 | |
AK127324.1 | 12 | 1539 | ||
AK222756.1 | 13 | 1563 | BAD96476.1 | |
AK294905.1 | 14 | 1698 | ||
AK297260.1 | ||||
AK299648.1 | ||||
AK301514.1 | 11 | 1229 | ||
AK304264.1 | 12 | 1469 | ||
AK307624.1 | 11 | 1152 | ||
AK314178.1 | 13 | 1561 | ||
BC004560.2 | AAH04560.1 | |||
BC017726.1 | 13 | 1488 | ||
BC072436.1 | 13 | 1536 | AAH72436.1 | |
DQ350134.1 | 13 | 1558 | ABC75106.1 | |
DQ372932.1 | 13 | 1558 | ABD14370.1 | |
U65785.1 | 13 | 1538 | AAC50947.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |