Assay Details
Target Gene Details
Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573744 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss | FANCA TCF25 SPIRE2 LOC107984817 LOC105371419 ZNF276 |
esv2758671 | Chr.16:89804324 - 89901541 on Build GRCh38 | Loss | FANCA TCF25 SPIRE2 LOC107984817 LOC105371419 |
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | FANCA TUBB3 SPIRE2 LOC105371419 CENPBD1 CHMP1A VPS9D1 CDK10 SPATA2L TCF25 LOC107984817 SPATA33 GAS8-AS1 ZNF276 VPS9D1-AS1 DBNDD1 GAS8 MC1R AFG3L1P DEF8 DPEP1 |
nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss | TCF25 TUBB3 SPIRE2 GAS8-AS1 CENPBD1 DBNDD1 GAS8 MC1R AFG3L1P DEF8 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | FANCA TUBB8P7 RPL13 FAM157C SPG7 TCF25 LOC107984817 URAHP SPATA33 LOC100287036 DBNDD1 MC1R AFG3L1P DEF8 DPEP1 ANKRD11 TUBB3 SPIRE2 LOC101927817 LOC105371419 SNORD68 ZNF778 CENPBD1 CHMP1A LOC105371414 VPS9D1 CDK10 SPATA2L GAS8-AS1 ZNF276 PRDM7 VPS9D1-AS1 GAS8 CPNE7 |
nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain | TCF25 TUBB3 SPIRE2 LOC105371419 CENPBD1 DBNDD1 GAS8 MC1R AFG3L1P DEF8 |
nsv1066499 | Chr.16:89891957 - 90028891 on Build GRCh38 | Loss | TCF25 TUBB3 CENPBD1 DBNDD1 GAS8 MC1R AFG3L1P DEF8 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | GALNS PABPN1L LOC101927793 LOC105371409 TCF25 LOC107984817 SLC22A31 DBNDD1 MC1R DEF8 DPEP1 IL17C ANKRD11 LOC100289580 SNORD68 CDT1 ZNF778 CHMP1A LOC107987238 VPS9D1 CDK10 SPATA2L APRT GAS8-AS1 ZNF276 PRDM7 GAS8 RNF166 CPNE7 PIEZO1 ACSF3 CBFA2T3 FANCA TUBB8P7 CDH15 RPL13 CTU2 FAM157C MIR4722 SPG7 CYBA URAHP SPATA33 SNAI3 LOC100287036 AFG3L1P SNAI3-AS1 LINC00304 TRAPPC2L TUBB3 SPIRE2 LOC101927817 LOC105371419 CENPBD1 LOC100129697 LOC105371414 LOC400558 VPS9D1-AS1 LOC339059 MVD |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |