Assay Details
Target Gene Details
Entrez Gene ID: | 60343 |
Gene Name: | family with sequence similarity 3 member A |
Gene Aliases: |
2.19, DLD, DXS560S, XAP-7 |
Location: |
Chr.X:154506159-154516249 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | FAM223A SLC10A3 MIR1184-2 F8A3 OPN1LW ATP6AP1 CMC4 TKTL1 DKC1 HCFC1 HCFC1-AS1 CTAG2 TEX28 SMIM9 TAZ FUNDC2 TMLHE-AS1 H2AFB2 PLXNA3 BRCC3 MPP1 MIR6858 IRAK1 EMD TMLHE GDI1 RPL10 RAB39B IKBKG CTAG1A G6PD MIR664B CTAG1B OPN1MW2 MECP2 DNASE1L1 CLIC2 OPN1MW3 LAGE3 SPRY3 LOC105373386 H2AFB1 H2AFB3 SNORA56 LOC107987332 MIR1184-3 FAM223B FLNA VBP1 TMEM187 MIR3202-1 FAM3A MTCP1 OPN1MW MIR1184-1 GAB3 F8A1 LOC101927830 F8A2 F8 MIR3202-2 MIR718 CH17-340M24.3 SNORA70 SNORA36A FAM50A UBL4A |
nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain | FAM223A OPN1MW2 MECP2 DNASE1L1 SLC10A3 OPN1MW3 LAGE3 LOC105373386 OPN1LW ATP6AP1 LOC107987332 TKTL1 FLNA TEX28 TAZ PLXNA3 FAM3A OPN1MW MIR6858 EMD GDI1 RPL10 IKBKG CTAG1A CH17-340M24.3 SNORA70 G6PD FAM50A UBL4A |
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | BGN ATP2B3 FAM223A AVPR2 DUSP9 ZFP92 SLC10A3 BCAP31 RENBP OPN1LW ATP6AP1 TREX2 PNMA6A TKTL1 HCFC1 ZNF275 PLXNB3 HCFC1-AS1 IDH3G FAM58A TEX28 TAZ NAA10 LOC105373381 PLXNA3 PDZD4 MIR6858 IRAK1 EMD GDI1 RPL10 SLC6A8 IKBKG CTAG1A G6PD SSR4 LOC107983954 ARHGAP4 OPN1MW2 MECP2 MAGEA1 DNASE1L1 LCA10 OPN1MW3 ABCD1 LAGE3 LOC105373386 SRPK3 L1CAM LOC107987332 HAUS7 PNCK FLNA TMEM187 MIR3202-1 FAM3A OPN1MW MIR3202-2 MIR718 CH17-340M24.3 SNORA70 LOC105373378 FAM50A LOC105373383 LOC105373377 UBL4A |
nsv508816 | Chr.X:154492171 - 154591783 on Build GRCh38 | Insertion | FAM223A FAM3A IKBKG CTAG1A G6PD |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |