Assay Details
Target Gene Details
Entrez Gene ID: | 64699 |
Gene Name: | transmembrane protease, serine 3 |
Gene Aliases: |
DFNB10, DFNB8, ECHOS1, TADG12 |
Location: |
Chr.21:42371887-42396846 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS3 | NM_001256317.1 | 11 | 1308 | NP_001243246.1 |
NM_024022.2 | 11 | 1311 | NP_076927.1 | |
NM_032404.2 | 8 | 1009 | NP_115780.1 | |
NR_046020.1 | 11 | 2066 | ||
AB038157.1 | 11 | 1311 | BAB20077.1 | |
AB038158.1 | 9 | 1397 | BAB20078.1 | |
AB038159.1 | 8 | 978 | BAB20079.1 | |
AF201380.1 | 11 | 1253 | AAG37012.1 | |
AK131212.1 | 3 | 1842 | ||
AK172842.1 | 11 | 1271 | BAD18806.1 | |
AY358458.1 | 11 | 1232 | AAQ88823.1 | |
AY633572.1 | 11 | 2066 | AAT66641.1 | |
BC074846.2 | 10 | 1136 | AAH74846.1 | |
BC074847.2 | 10 | 1139 | AAH74847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1136042 | Chr.21:42374686 - 42377623 on Build GRCh38 | Deletion | TMPRSS3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |