Assay Details
Target Gene Details
Entrez Gene ID: | 10075 |
Gene Name: | HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase |
Gene Aliases: |
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1 |
Location: |
Chr.X:53532096-53686729 on Build GRCh38 |
Assay Gene Location: | Within Exon 91 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | MAGEH1 MIR6857 GPR173 WNK3 GNL3L TSPYL2 FAM120C PFKFB1 PAGE3 FAM104B PAGE2B KDM5C MIR6895 LOC100421746 TSR2 FOXR2 MIRLET7F2 RIBC1 FGD1 MTRNR2L10 MIR6894 XAGE3 IQSEC2 FAM156B ALAS2 SMC1A MIR4536-1 PAGE2 USP51 HSD17B10 PAGE5 ITIH6 SNORA11 TRO FAM156A MIR98 APEX2 KANTR HUWE1 PHF8 MAGED2 SNORA109 MIR4536-2 |
esv3576879 | Chr.X:53529972 - 53593214 on Build GRCh38 | Gain | MIRLET7F2 MIR98 HUWE1 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | MAGEH1 MIR6857 GPR173 WNK3 GNL3L TSPYL2 FAM120C PFKFB1 PAGE3 FAM104B PAGE2B KDM5C MIR6895 LOC100421746 TSR2 FOXR2 MIRLET7F2 RIBC1 FGD1 MTRNR2L10 MIR6894 XAGE3 IQSEC2 FAM156B ALAS2 SMC1A MIR4536-1 PAGE2 USP51 HSD17B10 PAGE5 ITIH6 SNORA11 TRO FAM156A MIR98 APEX2 KANTR HUWE1 PHF8 MAGED2 SNORA109 MIR4536-2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |