Assay Details
Target Gene Details
Entrez Gene ID: | 7170 |
Gene Name: | tropomyosin 3 |
Gene Aliases: |
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30 |
Location: |
Chr.1:154155304-154192135 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPM3 | NM_001043351.1 | NP_001036816.1 | ||
NM_001043352.1 | 8 | 880 | NP_001036817.1 | |
NM_001043353.1 | 8 | 880 | NP_001036818.1 | |
NM_001278188.1 | NP_001265117.1 | |||
NM_001278189.1 | NP_001265118.1 | |||
NM_001278190.1 | NP_001265119.1 | |||
NM_001278191.1 | NP_001265120.1 | |||
NM_153649.3 | NP_705935.1 | |||
NR_103461.1 | ||||
AB062125.1 | BAB93457.1 | |||
AK026559.1 | ||||
AK056889.1 | ||||
AK056921.1 | ||||
AK092712.1 | BAC03956.1 | |||
AK123571.1 | ||||
AK125367.1 | ||||
AK298678.1 | ||||
AK301674.1 | ||||
AK308007.1 | ||||
AK315504.1 | ||||
AL537561.3 | ||||
AY004867.1 | 8 | 761 | AAF87083.1 | |
BC000771.2 | AAH00771.1 | |||
BC015403.1 | AAH15403.1 | |||
BC072428.1 | AAH72428.1 | |||
BF967161.1 | ||||
BI546300.1 | 8 | 793 | ||
BM674269.1 | 1 | 369 | ||
BQ933569.1 | ||||
BU956481.1 | ||||
BX419658.2 | 8 | 792 | ||
BX641075.1 | 1 | 2195 | ||
BX648485.1 | CAI46046.1 | |||
CR997359.1 | ||||
FJ224335.1 | ||||
KU178368.1 | ||||
KU178369.1 | ||||
KU178370.1 | ||||
X04588.1 | CAB37185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520398 | Chr.1:154095720 - 154169432 on Build GRCh38 | Gain+Loss | MIR5698 NUP210L TPM3 |
nsv1000153 | Chr.1:154101773 - 154179941 on Build GRCh38 | Gain | MIR5698 NUP210L TPM3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |