Assay Details
Target Gene Details
Entrez Gene ID: | 7852 |
Gene Name: | C-X-C motif chemokine receptor 4 |
Gene Aliases: |
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS |
Location: |
Chr.2:136114349-136118155 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCR4 | NM_003467.2 | NP_003458.1 | ||
AF348491.1 | AAK29630.1 | |||
AK129916.1 | ||||
AK296674.1 | ||||
AK312244.1 | ||||
BC020968.2 | AAH20968.1 | |||
D10924.1 | BAA01722.1 | |||
D28433.1 | BAA05799.1 | |||
L01639.1 | AAA16594.1 | |||
L06797.1 | AAA03209.1 | |||
M99293.1 | AAA16617.1 | |||
X71635.1 | CAA50641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523997 | Chr.2:136110306 - 136463173 on Build GRCh38 | Gain | CXCR4 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |