Assay Details
Target Gene Details
Entrez Gene ID: | 26261 |
Gene Name: | F-box protein 24 |
Gene Aliases: |
FBX24 |
Location: |
Chr.7:100582490-100601117 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXO24 | NM_001163499.1 | NP_001156971.1 | ||
NM_012172.4 | NP_036304.2 | |||
NM_033506.2 | NP_277041.1 | |||
XM_005250259.3 | XP_005250316.1 | |||
XM_011516022.1 | XP_011514324.1 | |||
XM_011516023.2 | XP_011514325.1 | |||
XM_017011961.1 | XP_016867450.1 | |||
AK292240.1 | ||||
AK301864.1 | ||||
AK302252.1 | ||||
AL136811.1 | CAB66745.1 | |||
BC067293.1 | AAH67293.1 |
Target Gene Details
Entrez Gene ID: | 100129845 |
Gene Name: | PCOLCE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:100589401-100604038 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCOLCE-AS1 | NR_038910.1 | |||
AK097705.1 | ||||
AK298216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)